AMBH580642FA Ambeed, Inc.

1-Phenyl-1H-imidazole-4-carbonitrile

  • MDL number MFCD21604049
  • Cas Number : 932974-43-1
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/932974-43-1.html

Safety Information

MSDS https://www.ambeed.com/products/932974-43-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart