AMBH580642FD Ambeed, Inc.

4-Chloro-2-methyl-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD15529557
  • Cas Number : 744209-64-1
  • Molecular Weight : 306.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-methyl-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C14H11ClN2O2S/c1-10-9-12-13(15)7-8-16-14(12)17(10)20(18,19)11-5-3-2-4-6-11/h2-9H,1H3
Inchi Key UJAMAALRPFEDPW-UHFFFAOYSA-N

Description

Synonym N/A
Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/744209-64-1.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/744209-64-1.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart