AMBH58064309 Ambeed, Inc.

5-Methoxy-1H-pyrazole

  • MDL number MFCD28011352
  • Cas Number : 215610-30-3
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Methoxy-1H-pyrazole
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/215610-30-3.html

Safety Information

signalword Warning
pcodes P261-P264-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/215610-30-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart