AMBH5806430E Ambeed, Inc.

GDC-0927

  • MDL number MFCD31631581
  • Cas Number : 1642297-01-5
  • Molecular Weight : 461.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1642297-01-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1642297-01-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart