AMBH58064310 Ambeed, Inc.

Methyl 7-amino-2,3-dihydrobenzofuran-4-carboxylate

  • MDL number MFCD24627745
  • Cas Number : 1280665-55-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 226.5
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 393.9±42.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1280665-55-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1280665-55-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart