AMBH58064316 Ambeed, Inc.

Decloxizine dihydrochloride

  • MDL number MFCD02183705
  • Cas Number : 13073-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/13073-96-6.html

Safety Information

MSDS https://www.ambeed.com/products/13073-96-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart