AMBH58064319 Ambeed, Inc.

Sodium ((2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)sulfamate

  • MDL number MFCD00057972
  • Cas Number : 38899-05-7
  • Molecular Weight : 281.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/38899-05-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/38899-05-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart