AMBH58064324 Ambeed, Inc.

1-Isopropyl-piperidine-4-carboxaldehyde

  • MDL number MFCD09971191
  • Cas Number : 280774-04-1
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/280774-04-1.html

Safety Information

MSDS https://www.ambeed.com/products/280774-04-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart