AMBH58064326 Ambeed, Inc.

cis-3-(Boc-amino)-4-(trifluormethyl)piperidine

  • MDL number MFCD22566226
  • Cas Number : 1363378-20-4
  • Molecular Weight : 268.2760

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1363378-20-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1363378-20-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart