AMBH5806432B Ambeed, Inc.

6-(Morpholinomethyl)pyridine-3-boronic Acid Pinacol Ester

  • MDL number MFCD16995640
  • Cas Number : 1160790-92-0
  • Molecular Weight : 304.1900

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1160790-92-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1160790-92-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart