AMBH5806432C Ambeed, Inc.

1,2,2-Trimethylpiperazine hydrochloride

  • MDL number MFCD09864551
  • Cas Number : 1312784-54-5
  • Molecular Weight : 164.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1312784-54-5.html

Safety Information

MSDS https://www.ambeed.com/products/1312784-54-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart