AMBH5806432D Ambeed, Inc.

4-Chloroisothiazolo[5,4-d]pyrimidine

  • MDL number MFCD23703348
  • Cas Number : 1483065-60-6
  • Molecular Weight : 171.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1483065-60-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1483065-60-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart