AMBH58064330 Ambeed, Inc.

Ecabet

  • MDL number MFCD11044526
  • Cas Number : 33159-27-2
  • Molecular Weight : 380.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/33159-27-2.html

Safety Information

MSDS https://www.ambeed.com/products/33159-27-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart