AMBH58064334 Ambeed, Inc.

2-Azabicyclo[2.2.2]octane

  • MDL number MFCD01664956
  • Cas Number : 280-38-6
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/280-38-6.html

Safety Information

MSDS https://www.ambeed.com/products/280-38-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart