AMBH58064335 Ambeed, Inc.

4-Chloro-5-iodo-2,6-dimethylpyrimidine

  • MDL number MFCD02081997
  • Cas Number : 83410-16-6
  • Molecular Weight : 268.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/83410-16-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/83410-16-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart