AMBH5806433A Ambeed, Inc.

2,4-Diphenyl-6-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,5-triazine

  • MDL number MFCD29918901
  • Cas Number : 1219956-23-6
  • Molecular Weight : 435.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1219956-23-6.html

Safety Information

MSDS https://www.ambeed.com/products/1219956-23-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart