AMBH58064349 Ambeed, Inc.

Methyl N-Phenylcarbamate

  • MDL number MFCD00027552
  • Cas Number : 2603-10-3
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 186.4�t 760 mmHg
Iupac Name methyl phenylcarbamate
Inchi Code 1S/C8H9NO2/c1-11-8(10)9-7-5-3-2-4-6-7/h2-6H,1H3,(H,9,10)
Inchi Key IAGUPODHENSJEZ-UHFFFAOYSA-N

Description

Synonym N/A
Purity 98%
Flash Point 66.5
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 186.4�t 760 mmHg
Product Link https://www.ambeed.com/products/2603-10-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2603-10-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart