AMBH58064359 Ambeed, Inc.

2,4-Dibromo-1-(trifluoromethoxy)benzene

  • MDL number MFCD20233323
  • Cas Number : 1840-97-7
  • Molecular Weight : 319.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1840-97-7.html

Safety Information

MSDS https://www.ambeed.com/products/1840-97-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart