AMBH58064360 Ambeed, Inc.

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

  • MDL number MFCD22494032
  • Cas Number : 873663-50-4
  • Molecular Weight : 234.1025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/873663-50-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/873663-50-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart