AMBH58064365 Ambeed, Inc.

Potassium triphosphate

  • MDL number MFCD00054129
  • Cas Number : 13845-36-8
  • Molecular Weight : 448.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name pentapotassium triphosphate
Inchi Code 1S/5K.H5O10P3/c;;;;;1-11(2,3)9-13(7,8)10-12(4,5)6/h;;;;;(H,7,8)(H2,1,2,3)(H2,4,5,6)/q5*+1;/p-5
Inchi Key ATGAWOHQWWULNK-UHFFFAOYSA-I

Description

Purity 96%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/13845-36-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13845-36-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart