AMBH5806436B Ambeed, Inc.

Ethyl 3-(4-methoxyphenyl)propanoate

  • MDL number MFCD02258632
  • Cas Number : 22767-72-2
  • Molecular Weight : 208.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/22767-72-2.html

Safety Information

MSDS https://www.ambeed.com/products/22767-72-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart