AMBH5806436E Ambeed, Inc.

4,4'-Bis(2,2-diphenylvinyl)-1,1'-biphenyl

  • MDL number MFCD03093280
  • Cas Number : 142289-08-5
  • Molecular Weight : 510.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/142289-08-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/142289-08-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart