AMBH5806436F Ambeed, Inc.

Potassium trifluoro(1H-pyrazol-4-yl)borate

  • MDL number MFCD09992891
  • Cas Number : 1111732-81-0
  • Molecular Weight : 173.9738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1111732-81-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1111732-81-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart