AMBH58064372 Ambeed, Inc.

2-Bromo-4-fluoro-5-methylpyridine

  • MDL number MFCD16609962
  • Cas Number : 1211537-29-9
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1211537-29-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1211537-29-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart