AMBH58064376 Ambeed, Inc.

2-(2-Thiazolyl)acetic Acid

  • MDL number MFCD02094168
  • Cas Number : 188937-16-8
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/188937-16-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/188937-16-8.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart