AMBH58064377 Ambeed, Inc.

9-Methylene-9H-fluorene

  • MDL number MFCD11850122
  • Cas Number : 4425-82-5
  • Molecular Weight : 178.2292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/4425-82-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4425-82-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart