AMBH5806437B Ambeed, Inc.

5-Fluoro-1H-indol-6-amine

  • MDL number MFCD12924060
  • Cas Number : 121716-63-0
  • Molecular Weight : 150.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/121716-63-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/121716-63-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart