AMBH5806437F Ambeed, Inc.

(S)-2-Amino-6-(3-carboxypropanamido)hexanoic acid

  • MDL number MFCD27924899
  • Cas Number : 52685-16-2
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/52685-16-2.html

Safety Information

MSDS https://www.ambeed.com/products/52685-16-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart