AMBH58064382 Ambeed, Inc.

5-(((6-Methoxypyridin-3-yl)amino)methylene)-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD00170987
  • Cas Number : 25063-69-8
  • Molecular Weight : 278.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/25063-69-8.html

Safety Information

MSDS https://www.ambeed.com/products/25063-69-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart