AMBH58064389 Ambeed, Inc.

tert-Butyl ((3R,4R)-4-hydroxypyrrolidin-3-yl)carbamate hydrochloride

  • MDL number MFCD28038712
  • Cas Number : 1820575-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1820575-70-9.html

Safety Information

MSDS https://www.ambeed.com/products/1820575-70-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart