AMBH58064394 Ambeed, Inc.

Bis(2-benzamidophenyl) Disulfide

  • MDL number MFCD00043806
  • Cas Number : 135-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/135-57-9.html

Safety Information

MSDS https://www.ambeed.com/products/135-57-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart