AMBH58064395 Ambeed, Inc.

2,3,4-Trichlorophenylboronic acid

  • MDL number MFCD03095112
  • Cas Number : 352530-21-3
  • Molecular Weight : 225.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/352530-21-3.html

Safety Information

MSDS https://www.ambeed.com/products/352530-21-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart