AMBH58064399 Ambeed, Inc.

3-Bromo-6-chloropyridin-2(1H)-one

  • MDL number MFCD22123755
  • Cas Number : 848423-85-8
  • Molecular Weight : 208.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-6-chloropyridin-2-ol
Purity 96
Flash Point 137.4
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 303.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/848423-85-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/848423-85-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart