AMBH580643A0 Ambeed, Inc.

Cinnamoyl chloride

  • MDL number MFCD00000732
  • Cas Number : 102-92-1
  • Molecular Weight : 166.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/102-92-1.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/102-92-1.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart