AMBH580643A3 Ambeed, Inc.

N-Pentylaniline

  • MDL number MFCD00059431
  • Cas Number : 2655-27-8
  • Molecular Weight : 163.2594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2655-27-8.html

Safety Information

signalword Warning
pcodes P261-P270-P280-P302+P352+P312+P362+P364-P304+P340+P312-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/2655-27-8.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332
Availability details for
Quantity
  • Availability
  • Add to Cart