AMBH580643A7 Ambeed, Inc.

1,2-Bis(4-(dimethylamino)phenyl)ethane-1,2-dione

  • MDL number MFCD00051389
  • Cas Number : 17078-27-2
  • Molecular Weight : 296.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/17078-27-2.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/17078-27-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart