AMBH580643A9 Ambeed, Inc.

Letermovir

  • MDL number MFCD22572725
  • Cas Number : 917389-32-3
  • Molecular Weight : 572.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/917389-32-3.html

Safety Information

MSDS https://www.ambeed.com/products/917389-32-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart