AMBH580643C4 Ambeed, Inc.

3,5-Dimethylisonicotinonitrile

  • MDL number MFCD11052634
  • Cas Number : 7584-08-9
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dimethylisonicotinonitrile
Inchi Code 1S/C8H8N2/c1-6-4-10-5-7(2)8(6)3-9/h4-5H,1-2H3
Inchi Key LXOPPFCWOVFQDR-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/7584-08-9.html

Safety Information

signalword Danger
pcodes P330-P233-P264-P280-P312-P270-P260-P337+P313-P305+P351+P338-P362-P403+P233-P261-P271-P405-P332+P313-P321-P304+P340
MSDS https://www.ambeed.com/products/7584-08-9.html
pictograms GHS06,GHS07
hstatements H301-H335-H319-H315
Availability details for
Quantity
  • Availability
  • Add to Cart