AMBH580643C6 Ambeed, Inc.

Eudesma-4,11-dien-3-one

  • MDL number MFCD09839027
  • Cas Number : 473-08-5
  • Molecular Weight : 218.3346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form Solid or liquid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/473-08-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/473-08-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart