AMBH580643CF Ambeed, Inc.

4-Nitro-5-(Trifluoromethyl)-1H-pyrazole

  • MDL number MFCD04971110
  • Cas Number : 1046462-99-0
  • Molecular Weight : 181.0728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1046462-99-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1046462-99-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart