AMBH580643D4 Ambeed, Inc.

5-(Bromomethyl)-2-chloropyridine

  • MDL number MFCD07368893
  • Cas Number : 182924-36-3
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/182924-36-3.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/182924-36-3.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart