AMBH580643D5 Ambeed, Inc.

4-Chloro-2-methyl-6-nitroquinazoline

  • MDL number MFCD11047030
  • Cas Number : 74151-22-7
  • Molecular Weight : 223.6158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/74151-22-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/74151-22-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart