AMBH580643D6 Ambeed, Inc.

N-Fmoc-3-nitro-L-tyrosine

  • MDL number MFCD00171383
  • Cas Number : 136590-09-5
  • Molecular Weight : 448.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/136590-09-5.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/136590-09-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart