AMBH580643D8 Ambeed, Inc.

10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride

  • MDL number MFCD00012314
  • Cas Number : 33948-19-5
  • Molecular Weight : 257.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-124�t N/A mmHg
Purity 98
Flash Point 194.2
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 397.5�t 760 mmHg
Product Link https://www.ambeed.com/products/33948-19-5.html

Safety Information

signalword Danger
pcodes P234-P260-P264-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310-P305+P351+P338+P310-P390-P405-P406-P501
hcodes 8
MSDS https://www.ambeed.com/products/33948-19-5.html
pictograms GHS05
hstatements H290-H314
Availability details for
Quantity
  • Availability
  • Add to Cart