AMBH580643DD Ambeed, Inc.

2,6-Dichloro-4-hydroxybenzaldehyde

  • MDL number MFCD06797919
  • Cas Number : 60964-09-2
  • Molecular Weight : 191.0115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/60964-09-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/60964-09-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart