AMBH580643DF Ambeed, Inc.

2-Methyl-3-phenylpropan-1-ol

  • MDL number MFCD20386075
  • Cas Number : 7384-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/7384-80-7.html

Safety Information

MSDS https://www.ambeed.com/products/7384-80-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart