AMBH580643ED Ambeed, Inc.

(S)-Benzyl 2-methylaziridine-1-carboxylate

  • MDL number MFCD19382510
  • Cas Number : 946088-68-2
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/946088-68-2.html

Safety Information

MSDS https://www.ambeed.com/products/946088-68-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart