AMBH580643F5 Ambeed, Inc.

6-Amino-3-methylpyrimidin-4(3H)-one

  • MDL number MFCD19204434
  • Cas Number : 1122-46-9
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-3-methyl-4(3H)-pyrimidinone
Inchi Code 1S/C5H7N3O/c1-8-3-7-4(6)2-5(8)9/h2-3H,6H2,1H3
Inchi Key KSRSEUOJMZPSFL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1122-46-9.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1122-46-9.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart