AMBH580643F8 Ambeed, Inc.

4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl

  • MDL number MFCD15146810
  • Cas Number : 147497-57-2
  • Molecular Weight : 265.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/147497-57-2.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/147497-57-2.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart