AMBH580643FA Ambeed, Inc.

Zosuquidar trihydrochloride

  • MDL number MFCD00942300
  • Cas Number : 167465-36-3
  • Molecular Weight : 636.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/167465-36-3.html

Safety Information

MSDS https://www.ambeed.com/products/167465-36-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart